Diethyl bromomalonate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Diethyl 2-bromomalonate

OR9987-25G OR9987-100G OR9987-500G
APOSOR9987-25GEA 22 EUR
APOSOR9987-25G APOSOR9987-100G APOSOR9987-500G
Diethyl bromomalonate 98%
Diethyl bromomalonate
Formule: C₇H₁₁BrO₄
MW: 239.07 g/mol
Kookpunt: 140…141 °C/30mmHg
Dichtheid: 1.402 g/cm³ (25 °C)
Vlampunt: 111 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009138
CAS nummer: 685-87-0
EINECS: 211-683-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR