3-Chloro-2-fluorobenzyl alcohol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0060-1G PC0060-5G
APOSPC0060-1GEA 29.2 EUR
APOSPC0060-1G APOSPC0060-5G
3-Chloro-2-fluorobenzyl alcohol 97%
3-Chloro-2-fluorobenzyl alcohol
Formule: C₇H₆ClFO
Smeltpunt: 39…41 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631549
CAS nummer: 261723-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR