3-Chloro-5-fluorobenzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0067-1G PC0067-25G PC0067-5G
APOSPC0067-1GEA 22 EUR
APOSPC0067-1G APOSPC0067-25G APOSPC0067-5G
3-Chloro-5-fluorobenzaldehyde 97%
3-Chloro-5-fluorobenzaldehyde
Formule: C₇H₄ClFO
Smeltpunt: 38…42 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02261734
CAS nummer: 90390-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR