2-Fluorophenylboronic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0073-100G PC0073-25G PC0073-500G
APOSPC0073-100GEA 120 EUR
APOSPC0073-100G APOSPC0073-25G APOSPC0073-500G
2-Fluorophenylboronic acid 97%
2-Fluorophenylboronic acid
Formule: C₆H₆BFO₂
Smeltpunt: 101…118 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00674013
CAS nummer: 1993-03-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR