5-Chloro-2-fluoroaniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0082-100G PC0082-25G PC0082-5G
APOSPC0082-100GEA 70.1 EUR
APOSPC0082-100G APOSPC0082-25G APOSPC0082-5G
5-Chloro-2-fluoroaniline 98%
5-Chloro-2-fluoroaniline
Formule: C₆H₅ClFN
Kookpunt: 102…105 °C/20mmHg
Smeltpunt: 20…22 °C
Dichtheid: 1,29 g/cm³ (25 °C)
Vlampunt: 103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00069416
CAS nummer: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR