2-Chloro-6-fluoro-3-methylbenzyl bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0114-10G PC0114-5G PC0114-1G
APOSPC0114-10GEA 316 EUR
APOSPC0114-10G APOSPC0114-5G APOSPC0114-1G
2-Chloro-6-fluoro-3-methylbenzyl bromide
2-Chloro-6-fluoro-3-methylbenzyl bromide
Formule: C₈H₇BrClF
Smeltpunt: 35…37 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631363
CAS nummer: 261762-87-2
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR