6-Chloro-2-fluoro-3-methylbenzyl bromide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0116-10G PC0116-1G PC0116-5G
APOSPC0116-10GEA 285 EUR
APOSPC0116-10G APOSPC0116-1G APOSPC0116-5G
6-Chloro-2-fluoro-3-methylbenzyl bromide 98%
6-Chloro-2-fluoro-3-methylbenzyl bromide
Formule: C₈H₇BrClF
Kookpunt: 130 °C/3mmHg
Smeltpunt: 52…58 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631373
CAS nummer: 261762-88-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR