5-Chloro-2-(trifluoromethyl)benzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0193-10G PC0193-1G PC0193-5G PC0193-25G
APOSPC0193-10GEA 409 EUR
APOSPC0193-10G APOSPC0193-1G APOSPC0193-5G APOSPC0193-25G
5-Chloro-2-(trifluoromethyl)benzyl alcohol
5-Chloro-2-(trifluoromethyl)benzyl alcohol
Formule: C₈H₆ClF₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631355
CAS nummer: 261763-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR