Pentafluorobenzyl chloride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0238-25G
APOSPC0238-25GEA 347 EUR
APOSPC0238-25G
Pentafluorobenzyl chloride 97%
Pentafluorobenzyl chloride
Formule: (C₆F₅)CH₂Cl
Kookpunt: 72 °C/22mmHg
Dichtheid: 1,59 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD17168377
CAS nummer: 653-35-0
EINECS: 211-501-0
UN: 2922
ADR: 8+6.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR