2-((Difluoromethyl)thio)benzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0270-1G
APOSPC0270-1GEA 46.8 EUR
APOSPC0270-1G
2-((Difluoromethyl)thio)benzoic acid 97%
2-((Difluoromethyl)thio)benzoic acid
Formule: C₈H₆F₂O₂S
Smeltpunt: 150…152 °C
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 79676-56-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR