4-Amino-2,6-difluorobenzotrifluoride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0289-5G
APOSPC0289-5GEA 555 EUR
APOSPC0289-5G
4-Amino-2,6-difluorobenzotrifluoride
4-Amino-2,6-difluorobenzotrifluoride
Formule: C₇H₄F₅N
Kookpunt: 103…105 °C
Smeltpunt: 66 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00190122
CAS nummer: 123950-44-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR