3-Chloro-5-fluorophenylacetonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0331-1G PC0331-5G PC0331-25G
APOSPC0331-1GEA 24.9 EUR
APOSPC0331-1G APOSPC0331-5G APOSPC0331-25G
3-Chloro-5-fluorophenylacetonitrile
3-Chloro-5-fluorophenylacetonitrile
Formule: C₈H₅ClFN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04038259
CAS nummer: 493038-93-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR