5-Fluoro-2-methylbenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0353-1G PC0353-5G PC0353-10G
APOSPC0353-1GEA 23.4 EUR
APOSPC0353-1G APOSPC0353-5G APOSPC0353-10G
5-Fluoro-2-methylbenzyl alcohol
5-Fluoro-2-methylbenzyl alcohol
Formule: C₈H₉FO
Smeltpunt: 36…40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631527
CAS nummer: 22062-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR