3-Fluoro-DL-phenylglycine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0377-1G PC0377-5G PC0377-25G
APOSPC0377-1GEA 43.8 EUR
APOSPC0377-1G APOSPC0377-5G APOSPC0377-25G
3-Fluoro-DL-phenylglycine
3-Fluoro-DL-phenylglycine
Formule: C₈H₈FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049325
CAS nummer: 7292-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR