3-Methyl-5-(trifluoromethyl)benzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0456-5G PC0456-10G PC0456-1G
APOSPC0456-5GEA 271 EUR
APOSPC0456-5G APOSPC0456-10G APOSPC0456-1G
3-Methyl-5-(trifluoromethyl)benzonitrile 97%
3-Methyl-5-(trifluoromethyl)benzonitrile
Formule: C₉H₆F₃N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631595
CAS nummer: 261952-04-9
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR