4-Methyl-3-(trifluoromethyl)benzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0478-5G PC0478-1G PC0478-250MG
APOSPC0478-5GEA 461 EUR
APOSPC0478-5G APOSPC0478-1G APOSPC0478-250MG
4-Methyl-3-(trifluoromethyl)benzyl alcohol
4-Methyl-3-(trifluoromethyl)benzyl alcohol
Formule: C₉H₉F₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631607
CAS nummer: 261952-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR