3-Methyl-5-(trifluoromethyl)benzyl bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC0483-1G PC0483-5G
APOSPC0483-1GEA 350 EUR
APOSPC0483-1G APOSPC0483-5G
3-Methyl-5-(trifluoromethyl)benzyl bromide
3-Methyl-5-(trifluoromethyl)benzyl bromide
Formule: C₉H₈BrF₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01631610
CAS nummer: 116070-36-1
UN: 3265
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR