2-(Trifluoromethoxy)anisole 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0515-1G PC0515-5G PC0515-25G
APOSPC0515-1GEA 22 EUR
APOSPC0515-1G APOSPC0515-5G APOSPC0515-25G
2-(Trifluoromethoxy)anisole 97%
2-(Trifluoromethoxy)anisole
Formule: C₈H₇F₃O₂
Kookpunt: 69…70 °C/10mmHg
Vlampunt: 47 °C
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 261952-22-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR