(1R)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC0586-1G PC0586-250MG
APOSPC0586-1GEA 503 EUR
APOSPC0586-1G APOSPC0586-250MG
(1R)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol 97%
(1R)-1-[2-(Trifluoromethyl)phenyl]ethan-1-ol
Formule: C₉H₉F₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03093000
CAS nummer: 127852-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR