2,4-Dibromo-6-fluorobenzoic acid ≥97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC0739-1G PC0739-5G PC0739-10G
APOSPC0739-1GEA 110 EUR
APOSPC0739-1G APOSPC0739-5G APOSPC0739-10G
2,4-Dibromo-6-fluorobenzoic acid ≥97%
2,4-Dibromo-6-fluorobenzoic acid
Formule: C₇H₃Br₂FO₂
MDL Nummer: MFCD03094087
CAS nummer: 183065-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR