Fmoc-D-4-fluorophe

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC0967-1G PC0967-25G PC0967-5G
APOSPC0967-1GEA 26.3 EUR
APOSPC0967-1G APOSPC0967-25G APOSPC0967-5G
Fmoc-D-4-fluorophe
Fmoc-D-4-fluorophe
Formule: C₂₄H₂₀FNO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00235900
CAS nummer: 177966-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR