2,4-Difluoro-5-nitrobenzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1001-25G PC1001-5G
APOSPC1001-25GEA 35.1 EUR
APOSPC1001-25G APOSPC1001-5G
2,4-Difluoro-5-nitrobenzonitrile 98%
2,4-Difluoro-5-nitrobenzonitrile
Formule: C₇H₂F₂N₂O₂
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07772030
CAS nummer: 67152-20-9
UN: 2811
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR