Allylpentafluorobenzene 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1006-25G PC1006-5G PC1006-1G
APOSPC1006-25GEA 139 EUR
APOSPC1006-25G APOSPC1006-5G APOSPC1006-1G
Allylpentafluorobenzene 96%
Allylpentafluorobenzene
Formule: (C₆F₅)CH₂CHCH₂
Kookpunt: 148…149 °C
Smeltpunt: –64 °C
Dichtheid: 1,358 g/cm³ (25 °C)
Vlampunt: 43 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000301
CAS nummer: 1736-60-3
EINECS: 217-083-6
UN: 1993
ADR: 3,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR