4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC10188-1G PC10188-500MG PC10188-5G
APOSPC10188-1GEA 173 EUR
APOSPC10188-1G APOSPC10188-500MG APOSPC10188-5G
4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile
4-(4-Chloro-3-fluorophenoxy)benzenecarbonitrile
Formule: C₁₃H₇ClFNO
Smeltpunt: 75…77 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01815095
CAS nummer: 449778-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR