3-Fluoro-5-(trifluoromethyl)phenylboronic acid 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC10381-1G PC10381-5G
APOSPC10381-1GEA 27.8 EUR
APOSPC10381-1G APOSPC10381-5G
3-Fluoro-5-(trifluoromethyl)phenylboronic acid 99%
3-Fluoro-5-(trifluoromethyl)phenylboronic acid
Formule: C₇H₅BF₄O₂
Smeltpunt: 178…180 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06657735
CAS nummer: 159020-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR