2-Nitro-4-[(trifluoromethyl)sulphonyl]aniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC10664-1G PC10664-5G
APOSPC10664-1GEA 110 EUR
APOSPC10664-1G APOSPC10664-5G
2-Nitro-4-[(trifluoromethyl)sulphonyl]aniline
2-Nitro-4-[(trifluoromethyl)sulphonyl]aniline
Formule: C₇H₅F₃N₂O₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00523745
CAS nummer: 400-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR