3-Iodo-4-(trifluoromethyl)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC10668-1G PC10668-5G PC10668-25G
APOSPC10668-1GEA 29.2 EUR
APOSPC10668-1G APOSPC10668-5G APOSPC10668-25G
3-Iodo-4-(trifluoromethyl)benzoic acid
3-Iodo-4-(trifluoromethyl)benzoic acid
Formule: C₈H₄F₃IO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD18398157
CAS nummer: 1034690-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR