(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulphonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC10692-1G
APOSPC10692-1GEA 461 EUR
APOSPC10692-1G
(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulphonate
(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulphonate
Formule: C₁₅H₅F₂₀IO₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00135120
CAS nummer: 77758-89-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR