(Triphenylphosphonio)difluoroacetate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC10699-5G
APOSPC10699-5GEA 103 EUR
APOSPC10699-5G
(Triphenylphosphonio)difluoroacetate 95%
(Triphenylphosphonio)difluoroacetate
Formule: C₂₀H₁₅F₂O₂P
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD28359690
CAS nummer: 1449521-05-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR