4-Fluoro-3-(trifluoromethyl)aniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1076-500G PC1076-100G PC1076-1KG
APOSPC1076-500GEA 176 EUR
APOSPC1076-500G APOSPC1076-100G APOSPC1076-1KG
4-Fluoro-3-(trifluoromethyl)aniline 98%
4-Fluoro-3-(trifluoromethyl)aniline
Formule: C₇H₅F₄N
Kookpunt: 207…208 °C
Dichtheid: 1,393 g/cm³ (25 °C)
Vlampunt: 92 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007834
CAS nummer: 2357-47-3
EINECS: 219-095-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR