4-Fluoro-2-methoxyphenylboronic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC11112-25G
APOSPC11112-25GEA 156 EUR
APOSPC11112-25G
4-Fluoro-2-methoxyphenylboronic acid 98%
4-Fluoro-2-methoxyphenylboronic acid
Formule: C₇H₈BFO₃
Smeltpunt: 131…136 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03788423
CAS nummer: 179899-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR