Benzenesulphonyl fluoride 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC1181-25G PC1181-5G
APOSPC1181-25GEA 438 EUR
APOSPC1181-25G APOSPC1181-5G
Benzenesulphonyl fluoride 99%
Benzenesulphonyl fluoride
Formule: C₆H₅FO₂S
MW: 160.17 g/mol
Kookpunt: 203…204 °C
Dichtheid: 1.333 g/cm³ (25 °C)
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007412
CAS nummer: 368-43-4
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR