4-(Trifluoromethyl)salicylic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1216-100G PC1216-25G PC1216-500G
APOSPC1216-100GEA 107 EUR
APOSPC1216-100G APOSPC1216-25G APOSPC1216-500G
4-(Trifluoromethyl)salicylic acid 98%
4-(Trifluoromethyl)salicylic acid
Formule: C₈H₅F₃O₃
Smeltpunt: 178…179 °C
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01529657
CAS nummer: 328-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR