trans-1,2-Bis(perfluorohexyl)ethylene 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1263-5G PC1263-1G
APOSPC1263-5GEA 234 EUR
APOSPC1263-5G APOSPC1263-1G
trans-1,2-Bis(perfluorohexyl)ethylene 99%
trans-1,2-Bis(perfluorohexyl)ethylene

Gas-carrier for biomedical applications, Jan Reiss et al., Nouveau Journal de Chimie, 8(4), 251, (1984).

Formule: C₁₄H₂F₂₆
Kookpunt: 195 °C
Dichtheid: 1,766 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094106
CAS nummer: 51249-67-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR