2-Bromo-α,α,α,3-tetrafluorotoluene 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1420T-1G PC1420T-25G PC1420T-5G
APOSPC1420T-1GEA 22 EUR
APOSPC1420T-1G APOSPC1420T-25G APOSPC1420T-5G
2-Bromo-α,α,α,3-tetrafluorotoluene 98%
2-Bromo-α,α,α,3-tetrafluorotoluene
Formule: C₇H₃BrF₄
Dichtheid: 1,741 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061243
CAS nummer: 104540-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR