1-Chloro-3-fluoro-2-nitrobenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1499-1G PC1499-25G PC1499-5G
APOSPC1499-1GEA 22 EUR
APOSPC1499-1G APOSPC1499-25G APOSPC1499-5G
1-Chloro-3-fluoro-2-nitrobenzene
1-Chloro-3-fluoro-2-nitrobenzene
Formule: C₆H₃ClFNO₂
Kookpunt: 148…152 °C/30mmHg
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06658264
CAS nummer: 64182-61-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR