Gatifloxacin

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC1528-100G PC1528-10G PC1528-25G
APOSPC1528-100GEA 249 EUR
APOSPC1528-100G APOSPC1528-10G APOSPC1528-25G
Gatifloxacin
Gatifloxacin
Formule: C₁₉H₂₂FN₃O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00895399
CAS nummer: 112811-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR