3-(Trifluoromethylthio)benzyl alcohol, Technical Grade

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC1529-1G PC1529-250MG PC1529-5G
APOSPC1529-1GEA 150 EUR
APOSPC1529-1G APOSPC1529-250MG APOSPC1529-5G
3-(Trifluoromethylthio)benzyl alcohol, Technical Grade
3-(Trifluoromethylthio)benzyl alcohol
Formule: C₈H₇F₃OS
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 82174-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR