2-Bromo-2,3,3,3-tetrafluoropropionyl chloride 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC1567P-250MG
APOSPC1567P-250MGEA 58.4 EUR
APOSPC1567P-250MG
2-Bromo-2,3,3,3-tetrafluoropropionyl chloride 96%
2-Bromo-2,3,3,3-tetrafluoropropionyl chloride
Formule: C₃BrClF₄O
Kookpunt: 67…70 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00153723
CAS nummer: 6066-45-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR