Potassium-4-(ethylthio)phenyltrifluoroborate 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC1733-10G PC1733-25G PC1733-5G
APOSPC1733-10GEA 264 EUR
APOSPC1733-10G APOSPC1733-25G APOSPC1733-5G
Potassium-4-(ethylthio)phenyltrifluoroborate 96%
Potassium-4-(ethylthio)phenyltrifluoroborate
Formule: C₈H₉BF₃KS
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115771
CAS nummer: 850623-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR