3-Nitrobenzenesulphonyl fluoride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC1787-25G PC1787-5G
APOSPC1787-25GEA 292 EUR
APOSPC1787-25G APOSPC1787-5G
3-Nitrobenzenesulphonyl fluoride 97%
3-Nitrobenzenesulphonyl fluoride
Formule: C₆H₄FNO₄S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00024845
CAS nummer: 349-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR