4-Chloro-3-(trifluoromethyl)phenol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC1930L-100G PC1930L-25G PC1930L-5G
APOSPC1930L-100GEA 84.7 EUR
APOSPC1930L-100G APOSPC1930L-25G APOSPC1930L-5G
4-Chloro-3-(trifluoromethyl)phenol 98%
4-Chloro-3-(trifluoromethyl)phenol
Formule: C₇H₄ClF₃O
Kookpunt: 120…122 °C/27mmHg
Smeltpunt: 42…46 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042525
CAS nummer: 6294-93-5
UN: 3261
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR