Methyl perfluoro-3,6-dioxaheptanoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC1947-25G PC1947-5G PC1947-100G
APOSPC1947-25GEA 190 EUR
APOSPC1947-25G APOSPC1947-5G APOSPC1947-100G
Methyl perfluoro-3,6-dioxaheptanoate
Methyl perfluoro-3,6-dioxaheptanoate
Formule: C₆H₃F₉O₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03094140
CAS nummer: 39187-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR