3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200043-1G PC200043-500MG PC200043-5G
APOSPC200043-1GEA 271 EUR
APOSPC200043-1G APOSPC200043-500MG APOSPC200043-5G
3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol
3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol
Formule: C₇H₉F₃N₂O
Smeltpunt: 79…81 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00794530
CAS nummer: 169213-78-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR