5-Bromo-6-(trifluoromethyl)-4-pyrimidinol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC200154-1G PC200154-5G PC200154-10G
APOSPC200154-1GEA 29.2 EUR
APOSPC200154-1G APOSPC200154-5G APOSPC200154-10G
5-Bromo-6-(trifluoromethyl)-4-pyrimidinol
5-Bromo-6-(trifluoromethyl)-4-pyrimidinol
Formule: C₅H₂BrF₃N₂O
Smeltpunt: 225…227 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09901776
CAS nummer: 942060-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR