5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC200161-1G PC200161-500MG
APOSPC200161-1GEA 212 EUR
APOSPC200161-1G APOSPC200161-500MG
5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
Formule: C₈H₃BrClF₃N₂
Smeltpunt: 74…76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11841008
CAS nummer: 1135283-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR