7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC200218-1G PC200218-500MG
APOSPC200218-1GEA 417 EUR
APOSPC200218-1G APOSPC200218-500MG
7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
7-Chloro-8-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine
Formule: C₈H₃ClF₃IN₂
Smeltpunt: 160…162 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD19982794
CAS nummer: 1331823-98-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR