2-Chloro-4-(4-fluorophenyl)-1,3,5-triazine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC200401-1G PC200401-5G
APOSPC200401-1GEA 184 EUR
APOSPC200401-1G APOSPC200401-5G
2-Chloro-4-(4-fluorophenyl)-1,3,5-triazine
2-Chloro-4-(4-fluorophenyl)-1,3,5-triazine
Formule: C₉H₅ClFN₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD11052420
CAS nummer: 1053658-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR