3-(4-Bromophenyl)-5-(trifluoromethyl)isoxazole-4-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC200553-1G
APOSPC200553-1GEA 673 EUR
APOSPC200553-1G
3-(4-Bromophenyl)-5-(trifluoromethyl)isoxazole-4-carboxylic acid
3-(4-Bromophenyl)-5-(trifluoromethyl)isoxazole-4-carboxylic acid
Formule: C₁₁H₅BrF₃NO₃
Smeltpunt: 145…148 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12196379
CAS nummer: 1159978-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR