5-Cyclopropyl-2-fluoropyridine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC201006-100MG PC201006-1G PC201006-250MG
APOSPC201006-100MGEA 169 EUR
APOSPC201006-100MG APOSPC201006-1G APOSPC201006-250MG
5-Cyclopropyl-2-fluoropyridine 97%
5-Cyclopropyl-2-fluoropyridine
Formule: C₈H₈FN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18384289
CAS nummer: 1034467-80-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR